Learn More
Selleck Chemical LLC Palomid 529 (P529) S2238-1g
SDP
Catalog No. 503112789
Encompass_Preferred
Palomid 529 (P529 SG 00529) inhibits both the mTORC1 and mTORC2 complexes reduces phosphorylation of pAktS473 pGSK3 S9 and pS6 Phase 1
Catalog No. 50-311-2789 Supplier Selleck Chemical LLC Supplier No. S22381G
Palomid 529 (P529 SG 00529) inhibits both the mTORC1 and mTORC2 complexes reduces phosphorylation of pAktS473 pGSK3 S9 and pS6 Phase 1