Learn More
Apexbio Technology LLC Palomid 529 914913-88-5 50mg
SDP
Catalog No. 5000094034
Encompass_Preferred
Palomid 529 (CAS 914913-88-5) is a small-molecule inhibitor of the PI3K/Akt/mTOR signaling pathway targeting both TORC1 and TORC2 complexes It exhibits potent anti-proliferative activity in the NCI-60 cell line panel with GI50 values below 35 M In endothelial cell assays Palomid 529 inhibits VEGF- and bFGF-induced proliferation with IC50 values of 20 nM and 30 nM respectively The PI3K/Akt/mTOR pathway is implicated in tumor cell survival proliferation angiogenesis and resistance to apoptosis Palomid 529 has also demonstrated the ability to suppress radiation-induced expression of Id-1 VEGF MMP-2 and MMP-9 expanding its utility in studies of cancer biology and radiotherapy sensitization
Catalog No. 50-000-94034 Supplier Apexbio Technology LLC Supplier No. A861850
Palomid 529 (CAS 914913-88-5) is a small-molecule inhibitor of the PI3K/Akt/mTOR signaling pathway targeting both TORC1 and TORC2 complexes It exhibits potent anti-proliferative activity in the NCI-60 cell line panel with GI50 values below 35 M In endothelial cell assays Palomid 529 inhibits VEGF- and bFGF-induced proliferation with IC50 values of 20 nM and 30 nM respectively The PI3K/Akt/mTOR pathway is implicated in tumor cell survival proliferation angiogenesis and resistance to apoptosis Palomid 529 has also demonstrated the ability to suppress radiation-induced expression of Id-1 VEGF MMP-2 and MMP-9 expanding its utility in studies of cancer biology and radiotherapy sensitization