Learn More
Medchemexpress LLC Palomid 529 | 914913-88-5 | 99.3% | C24H22O6 | 100 MG
SDP
Catalog No. 5000355491
Encompass_Preferred
Palomid 529 is a potent inhibitor of both mTORC1 and mTORC2 complexes, serving as a valuable reagent for various research applications. It interferes with the PI3K/AKT/mTOR pathway, making it suitable for studies in cellular signaling and apoptosis. This product is strictly for research use only and not intended for human or therapeutic applications.
  • Potent inhibitor of mTORC1 and mTORC2 complexes
  • Modulates PI3K/AKT/mTOR signaling pathways
  • Supports research in apoptosis
  • Intended solely for research purposes
Catalog No. 50-003-55491 Supplier Medchemexpress LLC Supplier No. HY14581100MG
Palomid 529 is a potent inhibitor of both mTORC1 and mTORC2 complexes, serving as a valuable reagent for various research applications. It interferes with the PI3K/AKT/mTOR pathway, making it suitable for studies in cellular signaling and apoptosis. This product is strictly for research use only and not intended for human or therapeutic applications.
  • Potent inhibitor of mTORC1 and mTORC2 complexes
  • Modulates PI3K/AKT/mTOR signaling pathways
  • Supports research in apoptosis
  • Intended solely for research purposes