Learn More
TARGETMOL CHEMICALS INC Palonosetron hydrochloride
SDP
Catalog No. 502880603
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Palonosetron hydrochloride (RS 25259) is a 5-HT3 antagonist used in the prevention and treatment of chemotherapy-induced nausea and vomiting. Purity 99.97%
Catalog No. 50-288-0603 Supplier TARGETMOL CHEMICALS INC Supplier No. T1156200MG
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Palonosetron hydrochloride (RS 25259) is a 5-HT3 antagonist used in the prevention and treatment of chemotherapy-induced nausea and vomiting. Purity 99.97%