Learn More
Cayman Chemical Palovaroten 10mg
SDP
Catalog No. 502938190
Encompass_Preferred
An orally bioavailable RARγ agonist; increases RARE activity and decreases IL-1β-induced NF-κB activity in primary mouse stromal cells at 60 nM; inhibits activin A-induced chondrogenic and osteogenic differentiation of mouse FAPs expressing ACVR1R206H at 10 nM; inhibits heterotopic ossification and preserves limb movement and growth in a ACVR1R206H FOP mouse model; decreases heterotopic ossification by 50 to 60% in a rat model of traumatic blast injury at 1 mg/kg per day for 14 days
Catalog No. 50-293-8190 Supplier Cayman Chemical Supplier No. 2846010 MG
An orally bioavailable RARγ agonist; increases RARE activity and decreases IL-1β-induced NF-κB activity in primary mouse stromal cells at 60 nM; inhibits activin A-induced chondrogenic and osteogenic differentiation of mouse FAPs expressing ACVR1R206H at 10 nM; inhibits heterotopic ossification and preserves limb movement and growth in a ACVR1R206H FOP mouse model; decreases heterotopic ossification by 50 to 60% in a rat model of traumatic blast injury at 1 mg/kg per day for 14 days