Learn More
Cayman Chemical PalustrIc AcId 10mg
SDP
Catalog No. 502918623
Encompass_Preferred
A diterpenoid with diverse biological activities; increases secreted hydrogen peroxide levels in isolated mouse macrophages at 2 mg/ml; toxic to water fleas and rainbow trout (LC50s = 0.1 and 0.5 mg/L, respectively); phloem levels are increased in mechanically injured, as well as mechanically injured and L. abietinum-infected, red pine and white spruce trees; has been found as an environmental contaminant in pulp and paper mill effluent
Catalog No. 50-291-8623 Supplier Cayman Chemical Supplier No. 3913610 MG
A diterpenoid with diverse biological activities; increases secreted hydrogen peroxide levels in isolated mouse macrophages at 2 mg/ml; toxic to water fleas and rainbow trout (LC50s = 0.1 and 0.5 mg/L, respectively); phloem levels are increased in mechanically injured, as well as mechanically injured and L. abietinum-infected, red pine and white spruce trees; has been found as an environmental contaminant in pulp and paper mill effluent