Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEGAComfort PAM™ ESD Anti-Static Insole

Catalog No. 18813501
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Men's: 8-9, Women's: 10-11
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-813-501 Men's: 8-9, Women's: 10-11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-813-501 Supplier MEGAComfort Supplier No. PAMESDM89/W1011
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Personal anti-fatigue mat ESD insole ergonomically designed by podiatrists, Dual layer 100% memory foam, Clinically proven field tested to reduce pain and fatigue, Max shock absorption

  • Personal Anti-Fatigue Mat (PAM™) ESD insole
  • Uses patented combination of high density dual layer 100% memory foam
  • For anti-static footwear only
  • ESD threads provide tested electro-static dissipation
  • Reduces pain and fatigue while increasing balance and comfort
  • Ideal for stationary and mobile workers
  • ESD anti-static threads provide electro-static dissipation
  • Silver ion anti-microbial technology in top cloth offering anti-bacterial, anti-perspiration and anti-odor
  • Machine washable

Note:

  • PAM™ ESD does not make regular footwear ESD compliant

Specifications

Color Black, Blue, Yellow
For Use With (Application) Stationary and mobile workers
For Use With (Equipment) Boots
Material Memory Foam, ESD Thread
Size Men's: 8-9, Women's: 10-11
Gender Unisex
Waterproof No
Product Type ESD Insole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.