Learn More
Invivogen PAM2CSK4 SYNT DIACYL LIPOP 1MG
SDP
Supplier:  Invivogen TLRLPM2S1
Encompass
Pam2CSK4 is a synthetic diacylated lipoprotein (LP). Current model: Diacylated LPs induce signaling through TLR2/6. Also reported: Induces signaling in a TLR6-independent manner.
Catalog No. NC0309196