Learn More
Medchemexpress LLC Pam2CSK4 (TFA) | 99.5% | 1271.82 (free base) | 5 MG
SDP
Catalog No. 5000422355
Encompass_Preferred
Pam2CSK4 (TFA) is a lipopeptide that acts as a TLR6-independent TLR2 ligand and agonist. It promotes platelet aggregation and increases platelet adhesion to collagen-coated surfaces in a TLR2/NF-κB/BTK-dependent manner. Pam2CSK4 (TFA) also activates iNOS expression and NO production in mouse macrophages.
  • Acts as a TLR6-independent TLR2 ligand and agonist.
  • Promotes platelet aggregation.
  • Increases platelet adhesion to collagen-coated surfaces.
  • Activates iNOS expression and NO production in mouse macrophages.
Catalog No. 50-004-22355 Supplier Medchemexpress LLC Supplier No. HYP1181A5MG
Pam2CSK4 (TFA) is a lipopeptide that acts as a TLR6-independent TLR2 ligand and agonist. It promotes platelet aggregation and increases platelet adhesion to collagen-coated surfaces in a TLR2/NF-κB/BTK-dependent manner. Pam2CSK4 (TFA) also activates iNOS expression and NO production in mouse macrophages.
  • Acts as a TLR6-independent TLR2 ligand and agonist.
  • Promotes platelet aggregation.
  • Increases platelet adhesion to collagen-coated surfaces.
  • Activates iNOS expression and NO production in mouse macrophages.