Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pam3-Cys-OH | 87420-41-5 | MFCD00153468 | 99.9% | 910.46 g/mol | C54H103NO7S | 50 MG
SDP

Catalog No. 5000283577
Encompass_Preferred

Pam3-Cys-OH is a lipoamino acid supplied as a white to off-white solid for biochemical research applications. It is a lipidated amino acid reagent with high lipophilicity and is commonly used as a biochemical assay reagent and research tool.

  • High purity suitable for research applications (listed as 99.89%).
  • Molecular formula C54H103NO7S.
  • Molecular weight 910.46 g/mol.
  • White to off-white solid appearance.
  • Soluble in DMSO at ~2 mg/mL; warming or ultrasonic assistance may aid dissolution.
  • Storage: powder at -20°C; in solution store at -80°C for long term.
  • For research use only; not for human therapeutic use.

Catalog No. 50-002-83577 Supplier Medchemexpress LLC Supplier No. HYW14178950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pam3-Cys-OH is a lipoamino acid supplied as a white to off-white solid for biochemical research applications. It is a lipidated amino acid reagent with high lipophilicity and is commonly used as a biochemical assay reagent and research tool.

  • High purity suitable for research applications (listed as 99.89%).
  • Molecular formula C54H103NO7S.
  • Molecular weight 910.46 g/mol.
  • White to off-white solid appearance.
  • Soluble in DMSO at ~2 mg/mL; warming or ultrasonic assistance may aid dissolution.
  • Storage: powder at -20°C; in solution store at -80°C for long term.
  • For research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.