Learn More
Invivogen Pam3CSK4 Synthetic triacylated lipopeptide; TLR2/TLR1 agonist, 1mg
SDP
Catalog No. NC9148916
Encompass
Pam3CSK4 TLR2 Ligands, Synthetic tripalmitoylated lipopeptide, Working Concentration: 100 to 300ng/mL, Quantity; 1mg
Catalog No. NC9148916 Supplier Invivogen Supplier No. TLRLPMS
Pam3CSK4 TLR2 Ligands, Synthetic tripalmitoylated lipopeptide, Working Concentration: 100 to 300ng/mL, Quantity; 1mg