Learn More
CPC Scientific Palmitoyl-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-Ser-Lys-Lys-Lys-Lys-OH (trifluoroacetate salt) 25MG
SDP
Catalog No. 502110071
Encompass
This peptide is a highly effective adjuvant for parenteral, oral and nasal immunizations. It readily dissolves in aqueous solvents and might also be applicable for DNA immunizations.SEQUENCE: Palmitoyl-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-Ser-Lys-Lys-Lys-Lys-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Palmitoyl-C((RS)-2,3-di(palmitoyloxy)-propyl)-SKKKKMOLECULAR FORMULA: C81H156N10O13S1MOLECULAR WEIGHT: 1510.26STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [112208-00-1]SYNONYMS: RESEARCH AREA: ImmunomodulationREFERENCES:J.Metzger et al., Int. J. Pept. Protein Res., 37, 46 (1991)U.v.d.Esche et al., Int. J. Immunopharmacol., 22, 1093 (2000)SKU(s): Pam4-001A, Pam4-001B
Catalog No. 50-211-0071 Supplier CPC Scientific Supplier No. PAM4001B
This peptide is a highly effective adjuvant for parenteral, oral and nasal immunizations. It readily dissolves in aqueous solvents and might also be applicable for DNA immunizations.SEQUENCE: Palmitoyl-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-Ser-Lys-Lys-Lys-Lys-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Palmitoyl-C((RS)-2,3-di(palmitoyloxy)-propyl)-SKKKKMOLECULAR FORMULA: C81H156N10O13S1MOLECULAR WEIGHT: 1510.26STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [112208-00-1]SYNONYMS: RESEARCH AREA: ImmunomodulationREFERENCES:J.Metzger et al., Int. J. Pept. Protein Res., 37, 46 (1991)U.v.d.Esche et al., Int. J. Immunopharmacol., 22, 1093 (2000)SKU(s): Pam4-001A, Pam4-001B