Learn More
Medchemexpress LLC HY-P1180A 1mg , Pam3CSK4 (TFA) CAS:112208-01-2 Purity:>98%
SDP
Catalog No. 502261985
Encompass_Preferred
HY-P1180A 1mg Pam3CSK4 (TFA) CAS: 112208-01-2 Pam3CSK4 TFA is a toll-like receptor 1/2 (TLR1/2) agonist with an EC50 of 0.47 ng/mL for human TLR1/2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1985 Supplier Medchemexpress LLC Supplier No. HYP1180A1MG
HY-P1180A 1mg Pam3CSK4 (TFA) CAS: 112208-01-2 Pam3CSK4 TFA is a toll-like receptor 1/2 (TLR1/2) agonist with an EC50 of 0.47 ng/mL for human TLR1/2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.