Learn More
AdipoGen Pam3Cys-Ser-Lys4 3HCl
SDP
Supplier:  AdipoGen AGCP30003M002
Encompass_Preferred
Chemical. CAS 112208-04-5. Formula C81H156N10O13S . 3HCl. MW 1510.3 . 109.4. Synthetic. Selective agonist of TLR1/TLR2 complex. Cell permeable, water soluble synthetic cationic lipohexapeptide analog of the immunologically active N-terminal portion of bacterial lipoprotein that potently activates monocytes and macrophages. Potent and effective immune adjuvant. Exerts strong local response, enhances IgG2a and IgG1 titers and upregulates proinflammatory and Th1 cytokine genes. Potent activator of the proinflammatory transcription factor NF-kappaB.
Catalog No. 50-256-2462