Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich PAMAM dendrimer

Catalog No. 41236825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-236-825G 25 g
41-236-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-236-825G Supplier Sigma Aldrich Supplier No. 41236825G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

PAMAM dendrimer is a poly(amidoamine) that is categorized under the class of dendritic polymers, which can be prepared by a two-step reaction that forms generations around the central ethylenediamine core. It consists of nitrogen and amide as functional groups that are structured in regular branched upon branched pattern. PAMAM is majorly used as a high capacity chelating agent.PAMAM dendrimer belongs to the family of dendrimers, which can be used in biomedical and drug delivery applications.Manufactured by Dendritech(R), Inc.

Specifications

CAS 155773-72-1
Density 0.854 g/mL (at 25°C)
Boiling Point 64.7°C
Molecular Formula C22H48N10O4
Refractive Index n20/D 1.364
Linear Formula NH2(CH2)2 NH2]:(G=0);dendri PAMAM(NH2)4
Quantity 25 g
MDL Number MFCD00192446
Molecular Weight (g/mol) 516.68
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.