Learn More
Medchemexpress LLC PAMAM dendrimer, ethylenediamine core, generation 3.0 | 153891-46-4 | 6908.84 g/mol | [NH2(CH2)2NH2]:(G=3); dendri PAMAM(NH2)32 | 250 MG
SDP
Catalog No. 5000201933
Encompass_Preferred
Polyamidoamine (PAMAM) is a generation-3 polyamidoamine dendrimer provided as a solution for formulation and drug-delivery research. It has internal encapsulation capability and a biocompatible surface suitable for nanoparticle and carrier studies.
  • Used for drug delivery and encapsulation research.
  • Provided as a 20% (w/w) solution in methanol for convenient handling.
  • Liquid appearance, colorless to light yellow.
  • Available in small research sizes for formulation development.
  • Contains PAMAM dendrimer structure (G3.0) with amine termini suitable for functionalization.
Catalog No. 50-002-01933 Supplier Medchemexpress LLC Supplier No. HY164657250MG
Polyamidoamine (PAMAM) is a generation-3 polyamidoamine dendrimer provided as a solution for formulation and drug-delivery research. It has internal encapsulation capability and a biocompatible surface suitable for nanoparticle and carrier studies.
  • Used for drug delivery and encapsulation research.
  • Provided as a 20% (w/w) solution in methanol for convenient handling.
  • Liquid appearance, colorless to light yellow.
  • Available in small research sizes for formulation development.
  • Contains PAMAM dendrimer structure (G3.0) with amine termini suitable for functionalization.