Learn More
Sigma Aldrich PAMAM dendrimer, ethylenediamine core, generation 3.5 solution
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Specifications
Specifications
| CAS | 192948-77-9 |
| Density | 0.81 g/mL (at 25°C) |
| Boiling Point | 65°C |
| Molecular Formula | C494H800N122Na64O188 |
| Refractive Index | n20/D 1.346 |
| Linear Formula | [NH2(CH2)2 NH2]:(G=3.5);dendri PAMAM(NHCH2CH2COONa)64 |
| Quantity | 10 g |
| MDL Number | MFCD00197936 |
| Synonym | PAMAM G3.5 |
| Molecular Weight (g/mol) | 12927.69 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.