Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich PAMAM dendrimer, ethylenediamine core, generation 4.5 solution

Catalog No. 47045725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-045-725G 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-045-725G Supplier Sigma Aldrich Supplier No. 4704572.5G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Manufactured by Dendritech(R), Inc.

Specifications

CAS 202009-66-3
Density 0.791 g/mL (at 25°C)
Boiling Point 65°C
Molecular Formula C1006H1632N250Na128O380
Refractive Index n20/D 1.337
Linear Formula [NH2(CH2)2 NH2]:(G=4.5);dendri PAMAM(NHCH2CH2COONa)128
Quantity 2.5 g
MDL Number MFCD00804247
Molecular Weight (g/mol) 26251.86
Biological Activity Eyes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.