Learn More
Cayman Chemical PAMAM Dendrimer G0 5 Carboxyl
SDP
Catalog No. 502918306
Encompass_Preferred
A carboxylate-terminated PAMAM dendrimer; conjugates of PAMAM G0.5 with the active fragment of oxaliplatin are cytotoxic to A2780, MCF-7, and Caco-2 cancer cells and BJ fibroblasts (IC50s = 0.03, 1.6, 0.18, and 3 µM, respectively)
Catalog No. 50-291-8306 Supplier Cayman Chemical Supplier No. 391041 G
A carboxylate-terminated PAMAM dendrimer; conjugates of PAMAM G0.5 with the active fragment of oxaliplatin are cytotoxic to A2780, MCF-7, and Caco-2 cancer cells and BJ fibroblasts (IC50s = 0.03, 1.6, 0.18, and 3 µM, respectively)