Learn More
Cayman Chemical PAMAM Dendrimer G1 5 Carboxyl
SDP
Catalog No. 502918240
Encompass_Preferred
A carboxylate-terminated PAMAM dendrimer; inhibits MERS-CoV-induced plaque formation in Vero cells at 10 µM; conjugates of PAMAM G1.5 with oxaliplatin are cytotoxic to A2780, MCF-7, and Caco-2 cancer cells and BJ fibroblasts (IC50s = 0.04, 1.6, 0.3, and 1.3 µM, respectively)
Catalog No. 50-291-8240 Supplier Cayman Chemical Supplier No. 391061 G
A carboxylate-terminated PAMAM dendrimer; inhibits MERS-CoV-induced plaque formation in Vero cells at 10 µM; conjugates of PAMAM G1.5 with oxaliplatin are cytotoxic to A2780, MCF-7, and Caco-2 cancer cells and BJ fibroblasts (IC50s = 0.04, 1.6, 0.3, and 1.3 µM, respectively)