Learn More
Cayman Chemical PAMAM Dendrimer G2 0 Amine w
SDP
Catalog No. 502918659
Encompass_Preferred
An amine-terminated PAMAM dendrimer; ~29 Å in diameter; has 16 surface groups; enhances the solubility and antibacterial activity of erythromycin against S. aureus when administered in combination with erythromycin at 15 mg/ml; increases the cell growth rate of HEK293T and HeLa cells from 100-500 nM but is cytotoxic to the same cells at ≥700 nM; enhances the nasal absorption of fluorescein isothiocyanate-labeled dextran in rats when administered in combination
Catalog No. 50-291-8659 Supplier Cayman Chemical Supplier No. 39072500 MG
An amine-terminated PAMAM dendrimer; ~29 Å in diameter; has 16 surface groups; enhances the solubility and antibacterial activity of erythromycin against S. aureus when administered in combination with erythromycin at 15 mg/ml; increases the cell growth rate of HEK293T and HeLa cells from 100-500 nM but is cytotoxic to the same cells at ≥700 nM; enhances the nasal absorption of fluorescein isothiocyanate-labeled dextran in rats when administered in combination