Learn More
Cayman Chemical PAMAM Dendrimer G2.0 Succinamic Acid (water solution), 1 g
SDP
Catalog No. 502918392
Encompass_Preferred
A succinamic acid-terminated PAMAM dendrimer; has 16 surface groups; an antagonist for anthrax toxin PA63 (IC50 = 879 nM) in lipid bilayers; active against S. aureus; inhibits plaque formation induced by MERS-CoV in Vero cells at 10 µM
Catalog No. 50-291-8392 Supplier Cayman Chemical Supplier No. 391191 G
A succinamic acid-terminated PAMAM dendrimer; has 16 surface groups; an antagonist for anthrax toxin PA63 (IC50 = 879 nM) in lipid bilayers; active against S. aureus; inhibits plaque formation induced by MERS-CoV in Vero cells at 10 µM