Learn More
Cayman Chemical PAMAM Dendrimer G3.0 Amine (water solution), CAS: 153891-46-4, 1 g
SDP
Catalog No. 502918660
Encompass_Preferred
An amine-terminated PAMAM dendrimer; ~36 Å in diameter; has 32 surface groups; cytotoxic to DU145 cells (IC50 = ~13.42 µM); toxic to zebrafish embryos, inducing mortality 72 hours post-fertilization at 0.2 µM; liposomes coated with PAMAM G3.0 and encapsulating berberine and chrysophanol reduce hydrogen peroxide-induced production of ROS in ARPE-19 cells and protect against retinal damage in a rat model of photooxidation-induced AMD
Catalog No. 50-291-8660 Supplier Cayman Chemical Supplier No. 390731 G
An amine-terminated PAMAM dendrimer; ~36 Å in diameter; has 32 surface groups; cytotoxic to DU145 cells (IC50 = ~13.42 µM); toxic to zebrafish embryos, inducing mortality 72 hours post-fertilization at 0.2 µM; liposomes coated with PAMAM G3.0 and encapsulating berberine and chrysophanol reduce hydrogen peroxide-induced production of ROS in ARPE-19 cells and protect against retinal damage in a rat model of photooxidation-induced AMD