Learn More
Cayman Chemical PAMAM Dendrimer G4.0 Amidoethanol (water solution), 1 g
SDP
Catalog No. 502918622
Encompass_Preferred
A hydroxyl-terminated PAMAM dendrimer; ~45 Å in diameter; has 64 surface groups; PAMAM G4.0 amidoethanol methylprednisolone conjugates decrease ovalbumin-induced increases in eosinophil numbers in BALF and lung tissue to a greater extent than methylprednisolone alone in a mouse model of lung inflammation; toxic to Anabaena and C. reinhardtii (EC90s = 5.13 and 1.02 mg/L, respectively)
Catalog No. 50-291-8622 Supplier Cayman Chemical Supplier No. 391131 G
A hydroxyl-terminated PAMAM dendrimer; ~45 Å in diameter; has 64 surface groups; PAMAM G4.0 amidoethanol methylprednisolone conjugates decrease ovalbumin-induced increases in eosinophil numbers in BALF and lung tissue to a greater extent than methylprednisolone alone in a mouse model of lung inflammation; toxic to Anabaena and C. reinhardtii (EC90s = 5.13 and 1.02 mg/L, respectively)