Learn More
Cayman Chemical PAMAM Dendrimer G4.5 Carboxylate (sodium salt) (water solution), CAS: 202009-66-3, 1 g
SDP
Catalog No. 502918664
Encompass_Preferred
A carboxylate-terminated PAMAM dendrimer; has been used in a drug delivery system; ~87.9 in diameter in water and has 96 surface groups; Unlike PAMAM G5.0, it does not rupture bilayer lipid membranes at 50 µM; PAMAM G4.5 carboxylate disulfur-linked aggregates encapsulating ferulic acid and paclitaxel induce apoptosis and decrease the mitochondrial membrane potential in multidrug-resistant KB-8-5 cancer cells at 10 µg/ml; complexes of PAMAM G4.5 carboxylate with the active component of cisplatin are cytotoxic to A2780 cancer cells (IC50 = 620 nM); a fluorescently labeled form of PAMAM G4.5 carboxylate selectively accumulates in tumors and kidneys over liver, spleen, heart, and lungs in a 4T1 murine mammary carcinoma model
Catalog No. 50-291-8664 Supplier Cayman Chemical Supplier No. 391091 G
A carboxylate-terminated PAMAM dendrimer; has been used in a drug delivery system; ~87.9 in diameter in water and has 96 surface groups; Unlike PAMAM G5.0, it does not rupture bilayer lipid membranes at 50 µM; PAMAM G4.5 carboxylate disulfur-linked aggregates encapsulating ferulic acid and paclitaxel induce apoptosis and decrease the mitochondrial membrane potential in multidrug-resistant KB-8-5 cancer cells at 10 µg/ml; complexes of PAMAM G4.5 carboxylate with the active component of cisplatin are cytotoxic to A2780 cancer cells (IC50 = 620 nM); a fluorescently labeled form of PAMAM G4.5 carboxylate selectively accumulates in tumors and kidneys over liver, spleen, heart, and lungs in a 4T1 murine mammary carcinoma model