Learn More
Cayman Chemical PAMAM Dendrimer G5 0 Amine w
SDP
Catalog No. 502918499
Encompass_Preferred
An amine-terminated PAMAM dendrimer; reduces viability, induces cell cycle arrest at the sub-G1/M phase, and increases LDH release from RAW 264.7 macrophages at 10 UG/ml; induces DNA fragmentation in RAW 264.7 cells at 15 UG/ml; PAMAM G5.0 in complex with siRNA targeting Pou5f1 mRNA, the gene encoding Oct4, reduces Oct4 expression in mouse embryonic stem cells; intravenous administration of PAMAM G5.0 in complex with reporter gene DNA increases reporter gene expression in the lung, spleen, and liver in mice; PAMAM G5.0-gadolinium conjugates increase small vessel contrast quality compared to DTPA-gadolinium conjugates
Catalog No. 50-291-8499 Supplier Cayman Chemical Supplier No. 39075500 MG
An amine-terminated PAMAM dendrimer; reduces viability, induces cell cycle arrest at the sub-G1/M phase, and increases LDH release from RAW 264.7 macrophages at 10 UG/ml; induces DNA fragmentation in RAW 264.7 cells at 15 UG/ml; PAMAM G5.0 in complex with siRNA targeting Pou5f1 mRNA, the gene encoding Oct4, reduces Oct4 expression in mouse embryonic stem cells; intravenous administration of PAMAM G5.0 in complex with reporter gene DNA increases reporter gene expression in the lung, spleen, and liver in mice; PAMAM G5.0-gadolinium conjugates increase small vessel contrast quality compared to DTPA-gadolinium conjugates