Learn More
Cayman Chemical PAMAM Dendrimer G6 0 Amine w
SDP
Catalog No. 502917098
Encompass_Preferred
An amine-terminated PAMAM dendrimer; has been used as a drug delivery system in vivo; ~67 Å in diameter and has 256 surface groups; active against clinical isolates of several bacteria (MICs = 0.25-25 UG/ml); increases the cell growth rate of HEK293T and HeLa cells from 100-500 nM but is cytotoxic to the same cells when used at ≥700 nM; increases the oral bioavailability of vardenafil in rabbits; has been used to deliver MRI contrast agents in mice
Catalog No. 50-291-7098 Supplier Cayman Chemical Supplier No. 39076100 MG
An amine-terminated PAMAM dendrimer; has been used as a drug delivery system in vivo; ~67 Å in diameter and has 256 surface groups; active against clinical isolates of several bacteria (MICs = 0.25-25 UG/ml); increases the cell growth rate of HEK293T and HeLa cells from 100-500 nM but is cytotoxic to the same cells when used at ≥700 nM; increases the oral bioavailability of vardenafil in rabbits; has been used to deliver MRI contrast agents in mice