Learn More
Cayman Chemical PAMAM Dendrimer G5.0 Amidoethanol (water solution), 500 mg
SDP
Catalog No. 502916940
Encompass_Preferred
A hydroxy-terminated PAMAM dendrimer; ~51 Å in diameter in PBS and has 128 surface groups; dendrimers encapsulating gold nanoparticles induce oxidation of carbon monoxide in a cell-free assay; binds to bilirubin (Kd = 8.5 µM); active against S. aureus; inhibits plaque formation induced by MERS-CoV in Vero cells at 10 µM; has been used in the formation and isolation of platinum nanoclusters for in vitro breast cancer subtyping
Catalog No. 50-291-6940 Supplier Cayman Chemical Supplier No. 39114500 MG
A hydroxy-terminated PAMAM dendrimer; ~51 Å in diameter in PBS and has 128 surface groups; dendrimers encapsulating gold nanoparticles induce oxidation of carbon monoxide in a cell-free assay; binds to bilirubin (Kd = 8.5 µM); active against S. aureus; inhibits plaque formation induced by MERS-CoV in Vero cells at 10 µM; has been used in the formation and isolation of platinum nanoclusters for in vitro breast cancer subtyping