Learn More
Cayman Chemical PAMAM Dendrimer G5.0 Succinamic Acid (water solution), 500 mg
SDP
Catalog No. 502918549
Encompass_Preferred
A succinamic acid-terminated PAMAM dendrimer; ~59 Å in diameter; has 128 surface groups; inhibits plaque formation induced by MERS-CoV in Vero cells at 10 µM; active against S. aureus; PAMAM G5.0-SAH encapsulating doxorubicin decreases cell viability in KB carcinoma cells; does not induce mortality in zebrafish at 250 ppm in tank water
Catalog No. 50-291-8549 Supplier Cayman Chemical Supplier No. 39122500 MG
A succinamic acid-terminated PAMAM dendrimer; ~59 Å in diameter; has 128 surface groups; inhibits plaque formation induced by MERS-CoV in Vero cells at 10 µM; active against S. aureus; PAMAM G5.0-SAH encapsulating doxorubicin decreases cell viability in KB carcinoma cells; does not induce mortality in zebrafish at 250 ppm in tank water