Learn More
Cayman Chemical PAMAM DendrImR G6 0 AmIdoeth
SDP
Catalog No. 502918666
Encompass_Preferred
A hydroxyl-terminated PAMAM dendrimer; ~65 Å in diameter in PBS; induces mortality in zebrafish embryos to a lesser extent than PAMAM dendrimer G6.0 amine but to a greater extent than PAMAM dendrimer G6.0 succinamic acid at 250 ppm in the tank water
Catalog No. 50-291-8666 Supplier Cayman Chemical Supplier No. 39115100 MG
A hydroxyl-terminated PAMAM dendrimer; ~65 Å in diameter in PBS; induces mortality in zebrafish embryos to a lesser extent than PAMAM dendrimer G6.0 amine but to a greater extent than PAMAM dendrimer G6.0 succinamic acid at 250 ppm in the tank water