Learn More
Cayman Chemical PAMAM Dendrimer G6.0 Amine (water solution), CAS: 163442-69-1, 50 mg
SDP
Catalog No. 502918360
Encompass_Preferred
An amine-terminated PAMAM dendrimer; has been used as a drug delivery system in vivo; ~67 Å in diameter and has 256 surface groups; active against clinical isolates of several bacteria (MICs = 0.25-25 µg/ml); increases the cell growth rate of HEK293T and HeLa cells from 100-500 nM but is cytotoxic to the same cells when used at ≥700 nM; increases the oral bioavailability of vardenafil in rabbits; has been used to deliver MRI contrast agents in mice
Catalog No. 50-291-8360 Supplier Cayman Chemical Supplier No. 3907650 MG
An amine-terminated PAMAM dendrimer; has been used as a drug delivery system in vivo; ~67 Å in diameter and has 256 surface groups; active against clinical isolates of several bacteria (MICs = 0.25-25 µg/ml); increases the cell growth rate of HEK293T and HeLa cells from 100-500 nM but is cytotoxic to the same cells when used at ≥700 nM; increases the oral bioavailability of vardenafil in rabbits; has been used to deliver MRI contrast agents in mice