Learn More
Cayman Chemical PAMAM DndrImR G6 0 AMDoth 50mg
SDP
Catalog No. 502917100
Encompass_Preferred
A hydroxyl-terminated PAMAM dendrimer; ~65 Å in diameter in PBS; induces mortality in zebrafish embryos to a lesser extent than PAMAM dendrimer G6.0 amine but to a greater extent than PAMAM dendrimer G6.0 succinamic acid at 250 ppm in the tank water
Catalog No. 50-291-7100 Supplier Cayman Chemical Supplier No. 3911550 MG
A hydroxyl-terminated PAMAM dendrimer; ~65 Å in diameter in PBS; induces mortality in zebrafish embryos to a lesser extent than PAMAM dendrimer G6.0 amine but to a greater extent than PAMAM dendrimer G6.0 succinamic acid at 250 ppm in the tank water