Learn More
Cayman Chemical PAMAM DndrImR G6 0 ScInam 50mg
SDP
Catalog No. 502916131
Encompass_Preferred
A succinamic acid-terminated PAMAM dendrimer; ~76 Å in diameter in PBS and has 256 surface groups; does not induce mortality in zebrafish embryos at 100 ppm in the tank water
Catalog No. 50-291-6131 Supplier Cayman Chemical Supplier No. 3912350 MG
A succinamic acid-terminated PAMAM dendrimer; ~76 Å in diameter in PBS and has 256 surface groups; does not induce mortality in zebrafish embryos at 100 ppm in the tank water