Learn More
Medchemexpress LLC Pamapimod | 449811-01-2 | >98.0% | 406.38 | C19H20F2N4O4 | 50 MG
SDP
Catalog No. 5000243937
Encompass_Preferred
Pamapimod is a selective p38 mitogen-activated protein kinase (MAPK) inhibitor used as a research compound to study MAPK signaling and inflammatory pathways. It is supplied for in vitro and in vivo experimental applications and is characterized in manufacturer and distributor datasheets for research use.
Catalog No. 50-002-43937 Supplier Medchemexpress LLC Supplier No. HY1040550MG
Pamapimod is a selective p38 mitogen-activated protein kinase (MAPK) inhibitor used as a research compound to study MAPK signaling and inflammatory pathways. It is supplied for in vitro and in vivo experimental applications and is characterized in manufacturer and distributor datasheets for research use.