Learn More
Medchemexpress LLC Pamicogrel | 101001-34-7 | 448.53 | 100 MG
SDP
Catalog No. 5000448599
Encompass_Preferred
Pamicogrel (KBT3022) is a cyclooxygenase (COX) inhibitor with platelet anti-aggregatory properties. It is currently under development by Kanebo for the treatment of chronic arterial occlusion and shows potential for both the prophylaxis and treatment of ischemic brain injury.
  • Acts as a cyclooxygenase (COX) inhibitor
  • Exhibits platelet anti-aggregatory properties
  • Developed for chronic arterial occlusion
  • Shows potential for prophylaxis of ischemic brain injury
  • Shows potential for treatment of ischemic brain injury
Catalog No. 50-004-48599 Supplier Medchemexpress LLC Supplier No. HYU00175100MG
Pamicogrel (KBT3022) is a cyclooxygenase (COX) inhibitor with platelet anti-aggregatory properties. It is currently under development by Kanebo for the treatment of chronic arterial occlusion and shows potential for both the prophylaxis and treatment of ischemic brain injury.
  • Acts as a cyclooxygenase (COX) inhibitor
  • Exhibits platelet anti-aggregatory properties
  • Developed for chronic arterial occlusion
  • Shows potential for prophylaxis of ischemic brain injury
  • Shows potential for treatment of ischemic brain injury