Learn More
Cayman Chemical Pamidronate (sodium salt), Purity: ≥98%, CAS: 57248-88-1, 100 mg
SDP
Catalog No. 502994142
Encompass_Preferred
An FPP synthase inhibitor (IC50s = 0.85 and 0.20 µM in J774 cells and for human recombinant FPP synthase, respectively); inhibits bone resorption in rats when used at doses of 16 µmol/kg per day and impairs long bone growth at ≥40 µmol/kg per day; has anti-hypercalcemic effects in a rat hypercalcemia model; high concentrations induce activation of MMP-2, NF-κB, and caspase-3, leading to cell death
Catalog No. 50-299-4142 Supplier Cayman Chemical Supplier No. 23699100 MG
An FPP synthase inhibitor (IC50s = 0.85 and 0.20 µM in J774 cells and for human recombinant FPP synthase, respectively); inhibits bone resorption in rats when used at doses of 16 µmol/kg per day and impairs long bone growth at ≥40 µmol/kg per day; has anti-hypercalcemic effects in a rat hypercalcemia model; high concentrations induce activation of MMP-2, NF-κB, and caspase-3, leading to cell death