Learn More
Medchemexpress LLC HY-104044 10mg , Pamiparib CAS:1446261-44-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10404410MG
Encompass_Preferred
HY-104044 10mg Pamiparib CAS: 1446261-44-4 Pamiparib (BGB-290) is an orally active, potent, highly selective PARP inhibitor, with IC50 values of 0.9 nM and 0.5 nM for PARP1 and PARP2, respectively. Pamiparib has potent PARP trapping, and capability to penetrate the brain, and can be used for the research of various cancers including the solid tumor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0460