Learn More
Selleck Chemical LLC Pamiparib
SDP
Catalog No. 5000000866
Encompass_Preferred
Pamiparib is a potent and selective inhibitor of PARP1 and PARP2 with IC50 values of 0 83 and 0 11 nM respectively in biochemical assays It shows high selectivity over other PARP enzymes
Catalog No. 50-000-00866 Supplier Selleck Chemical LLC Supplier No. S859225MG
Pamiparib is a potent and selective inhibitor of PARP1 and PARP2 with IC50 values of 0 83 and 0 11 nM respectively in biochemical assays It shows high selectivity over other PARP enzymes