Learn More
Cayman Chemical Pamoic Acid, Purity: ≥98%, CAS: 130-85-8, 50 g
SDP
Catalog No. 502955338
Encompass_Preferred
A potent agonist of GPR35 orphan receptors (EC50 = 79 nM in a β-arrestin recruitment assay); induces GPR35 translocation from the plasma membrane to the cytoplasm (EC50 =22 nM); increases ERK1/2 phosphorylation in a concentration-dependent manner in U20S cells expressing human GPR35a; has antinociceptive effects in a mouse abdominal constriction model of visceral pain perception (ED50 = 40.5 mg/kg)
Catalog No. 50-295-5338 Supplier Cayman Chemical Supplier No. 1394650 G
A potent agonist of GPR35 orphan receptors (EC50 = 79 nM in a β-arrestin recruitment assay); induces GPR35 translocation from the plasma membrane to the cytoplasm (EC50 =22 nM); increases ERK1/2 phosphorylation in a concentration-dependent manner in U20S cells expressing human GPR35a; has antinociceptive effects in a mouse abdominal constriction model of visceral pain perception (ED50 = 40.5 mg/kg)