Learn More
Medchemexpress LLC Pamoic acid | 130-85-8 | 99.69% | 388.38 | 5 G
SDP
Catalog No. 5000366586
Encompass_Preferred
Pamoic acid (Embonic acid) is a potent GPR35 agonist with an EC50 of 79 nM. It exhibits neuroprotective and anti-inflammatory properties.
  • Potent GPR35 agonist with an EC50 of 79 nM.
  • Exhibits neuroprotective properties.
  • Exhibits anti-inflammatory properties.
Catalog No. 50-003-66586 Supplier Medchemexpress LLC Supplier No. HYW0086135G
Pamoic acid (Embonic acid) is a potent GPR35 agonist with an EC50 of 79 nM. It exhibits neuroprotective and anti-inflammatory properties.
  • Potent GPR35 agonist with an EC50 of 79 nM.
  • Exhibits neuroprotective properties.
  • Exhibits anti-inflammatory properties.