Learn More
Medchemexpress LLC Pamoic acid | 130-85-8 | 99.7% | 388.38 | 1 ML
SDP
Catalog No. 5000452714
Encompass_Preferred
Pamoic acid (Embonic acid) is a potent GPR35 agonist with an EC50 of 79 nM. It exhibits neuroprotective and anti-inflammatory properties.
  • Potent GPR35 agonist
  • Exhibits neuroprotective properties
  • Exhibits anti-inflammatory properties
Catalog No. 50-004-52714 Supplier Medchemexpress LLC Supplier No. HYW00861310MM/1ML
Pamoic acid (Embonic acid) is a potent GPR35 agonist with an EC50 of 79 nM. It exhibits neuroprotective and anti-inflammatory properties.
  • Potent GPR35 agonist
  • Exhibits neuroprotective properties
  • Exhibits anti-inflammatory properties