Learn More
Medchemexpress LLC Pamoic acid | 130-85-8 | 99.7% | 388.38 | 25 G
SDP
Catalog No. 5000451410
Encompass_Preferred
Pamoic acid (Embonic acid) is a potent GPR35 agonist with an EC50 of 79 nM. It exhibits neuroprotective and anti-inflammatory properties.
  • Potent GPR35 agonist (EC50 of 79 nM).
  • Exhibits neuroprotective properties.
  • Exhibits anti-inflammatory properties.
  • Reduces infarct size in a mouse model of stroke.
  • Increases noninflammatory Ly-6CLo monocytes/macrophages and reduces neutrophil counts in the ischemic brain.
  • GPR35 activation may increase ERK1/2 phosphorylation, initiating an anti-inflammatory signal by suppressing NF-κB-dependent inflammatory genes.
Catalog No. 50-004-51410 Supplier Medchemexpress LLC Supplier No. 25GHYW008613
Pamoic acid (Embonic acid) is a potent GPR35 agonist with an EC50 of 79 nM. It exhibits neuroprotective and anti-inflammatory properties.
  • Potent GPR35 agonist (EC50 of 79 nM).
  • Exhibits neuroprotective properties.
  • Exhibits anti-inflammatory properties.
  • Reduces infarct size in a mouse model of stroke.
  • Increases noninflammatory Ly-6CLo monocytes/macrophages and reduces neutrophil counts in the ischemic brain.
  • GPR35 activation may increase ERK1/2 phosphorylation, initiating an anti-inflammatory signal by suppressing NF-κB-dependent inflammatory genes.