Learn More
Apexbio Technology LLC Pamoic acid disodium salt,50mg CAS# 6640-22-8
SDP
Catalog No. 502240550
Encompass_Preferred
Pamoic acid disodium salt, a supposedly inactive component in many formulations of drugs used to modulate release, is a potent agonist of the orphan receptor GPR35. GPR35 is a class A, rhodopsin-like G protein-coupled receptor (GPCR), strongly expressed in the lower intestine and colon, dorsal root ganglia, as well as a variety of immune cells including monocytes and dendritic cells. Targeting GPR35 provides potential therapeutic opportunities in a range of conditions, such as inflammation, pain and cancer. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0550 Supplier Apexbio Technology LLC Supplier No. B763850
Pamoic acid disodium salt, a supposedly inactive component in many formulations of drugs used to modulate release, is a potent agonist of the orphan receptor GPR35. GPR35 is a class A, rhodopsin-like G protein-coupled receptor (GPCR), strongly expressed in the lower intestine and colon, dorsal root ganglia, as well as a variety of immune cells including monocytes and dendritic cells. Targeting GPR35 provides potential therapeutic opportunities in a range of conditions, such as inflammation, pain and cancer. Other sizes are also available. Please inqury us for quote.