Learn More
Cayman Chemical PAMP-12 (human, mouse, rat, porcine, bovine) (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502914905
Encompass_Preferred
An endogenous fragment and MRGPRX2 agonist; inhibits forskolin-induced cAMP accumulation in CHO cells expressing human MRGPRX2 (EC50 = 57.2 nM); selectively induces calcium mobilization in CHO cells expressing MRGPRX2 (EC50 = 41 nM) over those expressing MRGPRX1, MRGPRX3, or MRGPRX4 at 1 µM; an antagonist of nAChRs that inhibits carbachol-induced catecholamine release (IC50 = 1.3 µM) and calcium and sodium influx (IC50s = 0.39 and 0.87 µM, respectively), but not histamine-induced catecholamine release or calcium and sodium influx (IC50s = >1 µM for all), in primary bovine adrenal chromaffin cells; reduces mean arterial blood pressure in normotensive rats at 10 to 50 nmol/kg
Catalog No. 50-291-4905 Supplier Cayman Chemical Supplier No. 3691010 MG
An endogenous fragment and MRGPRX2 agonist; inhibits forskolin-induced cAMP accumulation in CHO cells expressing human MRGPRX2 (EC50 = 57.2 nM); selectively induces calcium mobilization in CHO cells expressing MRGPRX2 (EC50 = 41 nM) over those expressing MRGPRX1, MRGPRX3, or MRGPRX4 at 1 µM; an antagonist of nAChRs that inhibits carbachol-induced catecholamine release (IC50 = 1.3 µM) and calcium and sodium influx (IC50s = 0.39 and 0.87 µM, respectively), but not histamine-induced catecholamine release or calcium and sodium influx (IC50s = >1 µM for all), in primary bovine adrenal chromaffin cells; reduces mean arterial blood pressure in normotensive rats at 10 to 50 nmol/kg