Learn More
Carbosynth LLC. PAMP (Human) (0.5 mg vial), 1ea, MW: 2460.8, Molecular formula: C112H178N36O27, CAS# 150238-87-2
SDP
Supplier:  Carbosynth LLC. PAM4291V0.5MG
Encompass_Preferred
Synonyms: Ala-Arg-Leu-Asp-Val-Ala-Ser-Glu-Phe-Arg-Lys-Lys-Trp-Asn-Lys-Trp-Ala-Leu-Ser-Arg-NH2 Proadrenomedullin N-terminal 20 Peptide (Human), Application: Hypotensive Peptide, Storage Temperature: -20°C, References: K. Kitamura, J. Sakata, K. Kangawa, M. Kojima, H. Matsuo, and T. Eto, Biochem. Biophys. Res. Commun., 194, 720 (1993). (Original; cDNA), H. Washimine, K. Kitamura, Y. Ichiki, Y. Yamamoto, K. Kangawa, H. Matsuo, and T. Eto, Biochem. Biophys. Res. Commun., 202, 1081 (1994). (Distribution in Human Tissue), K. Kitamura, K. Kangawa, Y. Ishiyama, H. Washimine, Y. Ichiki, M. Kawamoto, N. Minamino, H. Matsuo, and T. Eto, FEBS Lett., 351, 35 (1994). (Pharmacol.)
Catalog No. 50-168-6124