Learn More
Medchemexpress LLC Pamufetinib mesylate | 1688673-09-7 | 99.2% | 614.66 | C28H27FN4O7S2 | 25MG
SDP
Catalog No. 5000238520
Encompass_Preferred
Pamufetinib mesylate is a research-stage small-molecule kinase inhibitor that targets vascular endothelial growth factor receptors (VEGFR) and hepatocyte growth factor receptor (c-Met/HGFR). It exhibits low-nanomolar inhibition of recombinant VEGFR2 and MET and is used in preclinical studies of angiogenesis and MET-driven signaling in oncology research. The compound is provided as the mesylate salt for laboratory use and should be stored under controlled conditions to maintain stability.
  • Provides low-nanomolar inhibition of VEGFR2 and MET (IC50 ≈ 30-32 nM).
  • Suitable for preclinical studies of angiogenesis and MET-dependent signaling.
  • High chemical purity as confirmed by batch analysis (~99.2%).
  • Mesylate salt form for convenient laboratory handling.
  • Stable when stored under recommended conditions to preserve activity.
Catalog No. 50-002-38520 Supplier Medchemexpress LLC Supplier No. HY12423A25MG
Pamufetinib mesylate is a research-stage small-molecule kinase inhibitor that targets vascular endothelial growth factor receptors (VEGFR) and hepatocyte growth factor receptor (c-Met/HGFR). It exhibits low-nanomolar inhibition of recombinant VEGFR2 and MET and is used in preclinical studies of angiogenesis and MET-driven signaling in oncology research. The compound is provided as the mesylate salt for laboratory use and should be stored under controlled conditions to maintain stability.
  • Provides low-nanomolar inhibition of VEGFR2 and MET (IC50 ≈ 30-32 nM).
  • Suitable for preclinical studies of angiogenesis and MET-dependent signaling.
  • High chemical purity as confirmed by batch analysis (~99.2%).
  • Mesylate salt form for convenient laboratory handling.
  • Stable when stored under recommended conditions to preserve activity.