Learn More
Medchemexpress LLC Pamufetinib mesylate | 1688673-09-7 | 99.2% | 614.66 g·mol-1 | C28H27FN4O7S2 | 10MM 1ML
SDP
Catalog No. 5000236894
Encompass_Preferred
Pamufetinib mesylate is the methanesulfonate salt of a small-molecule kinase inhibitor that targets VEGFR and hepatocyte growth factor receptor (c-Met/HGFR). Supplied as a 10 mM solution in DMSO (1 mL) for in vitro research use, it demonstrates potent activity with reported IC50 values of 30 nM (rVEGFR2) and 32 nM (rMET). Molecular formula: C28H27FN4O7S2; molecular weight: 614.66 g·mol-1. Purity: 99.17%.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • Suitable for in vitro kinase inhibition assays.
  • Potent VEGFR and c-Met inhibitor with low-nanomolar IC50 values.
  • High chemical purity (99.17%).
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-36894 Supplier Medchemexpress LLC Supplier No. HY12423A10MM 1ML
Pamufetinib mesylate is the methanesulfonate salt of a small-molecule kinase inhibitor that targets VEGFR and hepatocyte growth factor receptor (c-Met/HGFR). Supplied as a 10 mM solution in DMSO (1 mL) for in vitro research use, it demonstrates potent activity with reported IC50 values of 30 nM (rVEGFR2) and 32 nM (rMET). Molecular formula: C28H27FN4O7S2; molecular weight: 614.66 g·mol-1. Purity: 99.17%.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • Suitable for in vitro kinase inhibition assays.
  • Potent VEGFR and c-Met inhibitor with low-nanomolar IC50 values.
  • High chemical purity (99.17%).
  • For research use only; not for human or veterinary use.