Learn More
Medchemexpress LLC Pamufetinib mesylate | 1688673-09-7 | 99.2% | 614.66 g/mol | C28H27FN4O7S2 | 100MG
SDP
Catalog No. 5000236824
Encompass_Preferred
Pamufetinib mesylate is an ATP-competitive, small-molecule inhibitor that targets vascular endothelial growth factor receptor (VEGFR) and hepatocyte growth factor receptor (c-Met/HGFR). Supplied as the mesylate salt for research use only, it exhibits potent in vitro kinase inhibition and is provided as a high-purity solid with recommended storage conditions to preserve stability.
  • Potent dual inhibition of VEGFR2 and MET (IC50 ≈ 30-32 nM).
  • High reported purity (99.2%).
  • Provided as a mesylate salt to improve handling and solubility for assays.
  • Available in a 100 mg pack size for research applications.
  • Solid, white to off-white appearance suitable for accurate weighing.
  • Storage recommendations: 4°C for powder; in solvent -80°C for long-term stability.
Catalog No. 50-002-36824 Supplier Medchemexpress LLC Supplier No. HY12423A100MG
Add to Cart
Add to Cart
Pamufetinib mesylate is an ATP-competitive, small-molecule inhibitor that targets vascular endothelial growth factor receptor (VEGFR) and hepatocyte growth factor receptor (c-Met/HGFR). Supplied as the mesylate salt for research use only, it exhibits potent in vitro kinase inhibition and is provided as a high-purity solid with recommended storage conditions to preserve stability.
  • Potent dual inhibition of VEGFR2 and MET (IC50 ≈ 30-32 nM).
  • High reported purity (99.2%).
  • Provided as a mesylate salt to improve handling and solubility for assays.
  • Available in a 100 mg pack size for research applications.
  • Solid, white to off-white appearance suitable for accurate weighing.
  • Storage recommendations: 4°C for powder; in solvent -80°C for long-term stability.