Learn More
PTM BIO LLC PAN ANTI-CROTONYLLYSINE ANTBDY
Catalog No. NC0729948
Encompass
The pan polyclonal anti-crotonyllysine antibody was produced by immunizing rabbits with crotonylated BSA. Antibodies were purified by crotonyllysine agarose affinity chromatography.
Catalog No. NC0729948 Supplier PTM BIO LLC Supplier No. PTM501
Add to Cart
Add to Cart
The pan polyclonal anti-crotonyllysine antibody was produced by immunizing rabbits with crotonylated BSA. Antibodies were purified by crotonyllysine agarose affinity chromatography.