Learn More
PTM BIO LLC Pan anti-succinyllysine antibody, 100 uL
Catalog No. NC0301551
Encompass
Anti-succinyllysine antibody detects proteins post-translationally modified with Succinylation at lysine residues. The antibody recognizes succinyl-lysine in a wide range of sequence contexts. The antibody does not cross-react with acetylated BSA and unmodified BSA.
Catalog No. NC0301551 Supplier PTM BIO LLC Supplier No. PTM401
Add to Cart
Add to Cart
Anti-succinyllysine antibody detects proteins post-translationally modified with Succinylation at lysine residues. The antibody recognizes succinyl-lysine in a wide range of sequence contexts. The antibody does not cross-react with acetylated BSA and unmodified BSA.